Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adaptin ε Mouse anti-Human, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612019

 View more versions of this product

Catalog No. BDB612019


Add to cart

Description

Description

Sorting of integral membrane proteins is mediated by vesicular trafficking between a variety of organelles. Two sorting signals are tyrosine-based and dileucine-based signals that interact with heterotetrameric adaptor protein complexes (AP-1, AP-2, AP-3, and AP-4), which are associated with the vesicle coats. These coatomers contain two large adaptin proteins (γ, α, σ, ε and β1, β2, βσ2, σ3, σ4 respectively). The AP-1 and AP-3 complexes are involved in protein sorting from the TGN and endosomes, while AP-2 adaptor complexes are involved in clathrin-mediated endocytosis. AP-4 is associated with non-clathrin coated vesicles in the region of the TGN. This localization is disrupted by brefeldin A, indicating that AP-4 membrane attachment is regulated by small GTPases. The µ4 subunit of the AP-4 complex interacts with tyrosine-based signals on LAMP-2 during targeting to the endosomal-lysosomal system. Thus, AP-4 is a less abundant AP complex that may be important for vesicle trafficking from the Golgi to the endosomal-lysosomal system.

Specifications

Specifications

Adaptin ε
32
Unconjugated
Affinity Purified
Human Adaptin ε aa. 685-793
150μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.