Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCK5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513893

Catalog No. PIPA513893

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ADCK5 is member of the AarF domain containing kinase family.
Specifications

Specifications

ADCK5
Polyclonal
PBS with 0.09% sodium azide
Q3MIX3, Q80V03
AarF domain containing kinase 5; ADCK5;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
203054, 268822
Western Blot
Unconjugated
ADCK5
Liquid
ADCK5
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human ADCK5
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit