Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCY2 Rabbit anti-Human, Mouse, Rabbit, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131190

Catalog No. PA131190


Add to cart

Description

Description

PA1-31190 detects ADCY2 from rat, human, mouse samples. PA1-31190 has been successfully used in Western blot applications. The PA1-31190 immunogen is: Synthetic peptide unique to ADCY2 protein was used to generate antibodies in rabbit. All peptides were amidated before conjugation. Store at 4°C short term. For extended storage aliquot and store at -20°C or below. Avoid freeze-thaw cycles.

This gene encodes a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex.
Specifications

Specifications

ADCY2
Polyclonal
Unconjugated
ADCY2
Liquid
ADCY2
Synthetic peptide unique to ADCY2 protein was used to generate antibodies in rabbit. All peptides were amidated before conjugation.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rabbit, Rat
Western Blot
0.75 mg/mL
PBS with 1% BSA and 0.02% sodium azide
Q08462, Q80TL1, P26769
AC2, FLJ16822, FLJ45092, HBAC2, KIAA1060, MGC133314
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100009201, 108, 210044, 81636
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit