Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCY2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512328

Catalog No. PIPA512328

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ADCY2 is a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate(cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex.
Specifications

Specifications

ADCY2
Polyclonal
PBS with 0.09% sodium azide
Q08462
Adenylate cyclase type 2; Adenylate cyclase type II; ATP pyrophosphate-lyase 2; Adenylyl cyclase 2; ADCY2; KIAA1060
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
108
Immunohistochemistry, Western Blot
Unconjugated
ADCY2
Liquid
ADCY2
KLH conjugated synthetic peptide between 940-969 amino acids from the C-terminal region of human ADCY2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit