Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AAV Rabbit anti-Virus, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14106

Catalog No. PA14106


Add to cart

Description

Description

PA1-4106 detects adeno-associated virus. PA1-4106 has been successfully used in Western blot procedures. The PA1-4106 immunogen is a mix of synthetic peptides corresponding to residues T(75) P W G Y F D F N R F H C(86) of major coat protein VP3 [AAV 2], residues E(278) G G T N L L Q F P H V(289) of capsid protein VP1 [AAV 6], and residues 277-288 of capsid AAV 3.

Adeno-associated virus (AAV) is a small virus which infects humans and some other primate species. AAV is not currently known to cause disease. The virus causes a very mild immune response, lending further support to its apparent lack of pathogenicity.
Specifications

Specifications

AAV
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
Virus
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
HADV; AdV; AAV; Adeno-Associated Virus; Lentivirus
Synthetic peptides corresponding to residues 75-86 of major coat protein VP3 [AAV 2], 278-289 of capsid protein VP1 [AAV 6], and 277-288 of capsid AAV 3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit