Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADORA2A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521273

Catalog No. PIPA521273


Add to cart

Description

Description

PA5-21273 targets Adenosine A2A-R in WB applications and shows reactivity with Human samples. The PA5-21273 immunogen is a synthetic peptide corresponding to a region within amino acids 338 and 412 of Adenosine A2a Receptor (Uniprot IDNo.P29274). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

ADORA2A
Polyclonal
Unconjugated
ADORA2A
Liquid
ADORA2A
Synthetic peptide corresponding to a region within amino acids 338 and 412 of Adenosine A2a Receptor (Uniprot IDNo.P29274)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
P29274
ADORA2, RDC8, hA2aR, adenosine A2 receptor, adenosine receptor A2a, adenosine receptor subtype A2a
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
135
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit