Learn More
MilliporeSigma™ anti-Adenosine A2b Receptor ; Polyclonal; ™ (Chemicon™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB1589P/DEL
Catalog No. 50-173-189
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Adenosine A2b receptor. Recognizes a band of approximately 50-52 kDa in several rat, mouse and human tissues (thymus, placenta, colon and small intestine). The antibody also recognizes the 35 kDa band in small intestine. Does not recognize the native receptors. No bands were detected in skeletal muscle or heart. The antibody will not recognize native protein in immunoprecipitation experiments.
Pierce et al. 1992; Tucker & Linden 1993
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
Adenosine Receptor A2b
Affinity Purified
Amino acid sequence corresponding to the 2nd extracellular domain (16 amino acids) of the human brain A2bR gene.
RUO
Primary
Human, Murine, Rat
Unconjugated
Rabbit
50μg
Neuroscience
Polyclonal