Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine Deaminase Rabbit anti-Bovine, HRP, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA127246

Catalog No. PA127246


Add to cart

Description

Description

PA1-27246 detects Adenosine Deaminase from bovine samples. PA1-27246 has been successfully used in ELISA, immunoprecipitation, and Western blot procedures. Predicted molecular weight: 104 kDa The PA1-27246 immunogen is Adenosine Deaminase from calf spleen. Store at 4C short term. For extended storage aliquot and store at -20C or below. Avoid Freeze-thaw cycles.

Specifications

Specifications

Adenosine Deaminase
Polyclonal
HRP
ADA
Liquid
Rabbit
IgG
1 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
280712
ELISA, Immunoprecipitation, Western Blot
10 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2
P56658
ADA
Adenosine Deaminase from calf spleen.
IgG fraction
RUO
Antibody
Polyclonal
Bovine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit