Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Adenosine Receptor A2, Clone: 7F6-G5-A2, ™ (Upstate™)
Supplier: MilliporeSigma™ 05717/DEL
Description
Rosin, D. L., et al(1998). Immunohistochemical localization of adenosine A2A receptors in the rat central nervous system.J Comp Neurol, 401: 163-86.
Flow Cytometry, Immunohistochemistry, Western Blotting
Specifications
Adenosine Receptor A2 | |
Unconjugated | |
6His-tagged full length human recombinant A2A receptor. Epitope mapped to SQPLPGER in the third intracellular loop. | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
200 μg | |
Neuroscience | |
Canine, Human, Mouse, Rabbit, Rat | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction