Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus Type 3 Rabbit anti-Virus, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522818

Catalog No. PIPA522818


Add to cart

Description

Description

Adenoviruses are DNA viruses generally widespread in nature that are frequently the cause of acute upper respiratory tract infections (i.e. common colds). 49 human adenovirus serotypes have been discovered and divided into six subgroups, A to E, based on observation. They are an important cause of respiratory illness, conjunctivitis, gastroenteritis, and have more recently been recognized as a potential cause of sexually transmitted diseases. Several types have oncogenic potential though most cause self-limiting febrile illnesses characterized by inflammation of conjunctivae and the respiratory tract. The virus can be isolated from the majority of tonsils/adenoids surgically removed, indicating latent infections. It is not known how long the virus can persist in the body, or whether it is capable of reactivation after long periods. In patients experiencing immunosuppression (e.g. AIDS) it can be reactivated, causing disease. Adenovirus vectors, particularly those based on Ad5, have shown great promise for the development of gene therapy protocols and have been used for clinical purpose.
Specifications

Specifications

Adenovirus Type 3
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Virus
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
HADV; AdV; Adeno Virus; HADV-3; AdV-3; Adeno Virus 3
Synthetic peptide within residues 250-270 of the fiber knob protein of Ad 3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit