Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenovirus Type 1 Monoclonal Antibody (1M13), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA17599
Description
MA1-7599 detects Adenovirus from virus samples. MA1-7599 has been successfully used in ELISA, Western blot and Immunofluorescence procedures. The MA1-7599 immunogen is a purified Ad1 virus.
Adenovirus Type 1 Monoclonal antibody specifically detects Adenovirus Type 1 in Virus samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Adenovirus Type 1 | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA, Immunocytochemistry, Western Blot | |
1M13 | |
Unconjugated | |
Adeno Virus; Adeno Virus 1; AdV; AdV-1; HADV; HADV-1 | |
Purified Ad1 virus. | |
100 μg | |
Primary | |
4°C short term, -80°C long term | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction