Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus Type 5 Hexon Goat anti-Virus, HRP, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA173054

Catalog No. PA173054


Add to cart

Description

Description

The PA1-73054 antibody reacts with Adenovirus hexon. PA1-73054 has been successfully used in ELISA applications. The PA1-73054 antibody was raised against Hexon from Adenovirus, type 5. Store at 4°C for up to 6 months. For longer storage, store at -20°C, Avoiding freeze/thaw cycles. Protect from light. NOTE: Sodium azide is a potent inhibitor of peroxidase!

Adenoviruses are DNA viruses generally widespread in nature that are frequently the cause of acute upper respiratory tract infections (i.e. common colds). 49 human adenovirus serotypes have been discovered and divided into six subgroups, A to E, based on observation. They are an important cause of respiratory illness, conjunctivitis, gastroenteritis, and have more recently been recognized as a potential cause of sexually transmitted diseases. Several types have oncogenic potential though most cause self-limiting febrile illnesses characterized by inflammation of conjunctivae and the respiratory tract. The virus can be isolated from the majority of tonsils/adenoids surgically removed, indicating latent infections. It is not known how long the virus can persist in the body, or whether it is capable of reactivation after long periods. In patients experiencing immunosuppression (e.g. AIDS) it can be reactivated, causing disease. Adenovirus vectors, particularly those based on Ad5, have shown great promise for the development of gene therapy protocols and have been used for clinical purpose. Adenoviruses capsids have three principal protein components: the hexon, the penton, and the fiber. Hexon protein is a major coat protein (240 hexons per particle). Each hexon capsomere is homotrimer of the hexon protein, a complex protein of over 900 residues.
Specifications

Specifications

Adenovirus Type 5 Hexon
Polyclonal
HRP
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Virus
ELISA
1-2 mg/mL
PBS with 10mg/mL BSA and 0.002% thimerosal; pH 7.2
HADV; AdV; Adeno Virus; HADV-5; AdV-5; Adeno Virus 5
Hexon from Adenovirus, type 5.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit