Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCY2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527396

Catalog No. PIPA527396


Add to cart

Description

Description

PA5-27396 targets Adenylate cyclase 2 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27396 immunogen is recombit fragment corresponding to a region within amino acids 420 and 597 of Human Adenylate cyclase 2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex.
Specifications

Specifications

ADCY2
Polyclonal
Unconjugated
ADCY2
Liquid
ADCY2
Recombit fragment corresponding to a region within amino acids 420 and 597 of Human Adenylate cyclase 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q08462
AC2, HBAC2, 3′,5′-cyclic AMP synthetase, ATP pyrophosphate-lyase 2, adenylate cyclase II, adenylate cyclase type 2, adenylate cyclase type II, adenylyl cyclase 2, type II adenylate cyclase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
108
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit