Learn More
anti-ADH4, Clone: 1D2, Abnova™
Mouse Monoclonal
Manufacturer:  Abnova Corporation H00000127M03
 View more versions of this product
Catalog No. 89-016-053
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
ADH4
Unlabeled
ADH4 (NP_000661, 52 a.a. ∽151 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26kDa.
50μg
Monoclonal
1D2
Murine
IgG2a, κ
Primary
Human