Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADH6 Mouse anti-Human, Clone: 4G4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA518359

Catalog No. PIMA518359


Add to cart

Description

Description

Peptide Sequence: GSKHLDLLYP TILGHEGAGI VESIGEGVST VKPGDKVITL FLPQCGECTS CLNSEGNFCI QFKQSKTQLM SDGTSRFTCK GKSIYHFGNT

This gene encodes class V alcohol dehydrogenase, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. This gene is expressed in the stomach as well as in the liver, and it contains a glucocorticoid response element upstream of its 5 ft. UTR, which is a steroid hormone receptor binding site. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

ADH6
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P28332
ADH-5, alcohol dehydrogenase 6, aldehyde reductase
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
130
ELISA, Western Blot
4G4
Unconjugated
ADH6
Liquid
ADH6
ADH6 (NP_000663, 55 a.a. ∽ 144 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit