Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADH6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526212

Catalog No. PIPA526212

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ADH6 encodes class V alcohol dehydrogenase, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. This protein is expressed in the stomach as well as in the liver, and it contains a glucocorticoid response element upstream of its 5 ft. UTR, which is a steroid hormone receptor binding site.
Specifications

Specifications

ADH6
Polyclonal
PBS with 0.09% sodium azide
P28332
ADH-5, alcohol dehydrogenase 6, aldehyde reductase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
130
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ADH6
Liquid
ADH6
KLH conjugated synthetic peptide between 209-240 amino acids from the central region of human ADH6
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit