Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP4 Chicken anti-Human, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA19577

Catalog No. 01-674-453


Add to cart

Description

Description

PA1-9577 detects ALBP in human samples. PA1-9577 has been successfully used in Western blot and ELISA procedures. By Western blot, PA1-9577 detects a 15 kDa protein representing ALBP. PA1-9577 immunogen is a synthetic peptide based on human ALBP protein.

FABP4 belongs to the fatty acid binding protein (FABP) family. FABPs contribute to the transport of fatty acids and other lipids in various cellular pathways. FABP4 is expressed in adipocytes, macrophages, monocyte-derived dendritic cells, and myoepithelial cells. It delivers long-chain fatty acids and retinoic acid to their cognate receptors in the nucleus.
Specifications

Specifications

FABP4
Polyclonal
Unconjugated
FABP4
Liquid
FABP4
Synthetic peptide based on human ALBP protein.
Affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
0.5 mg/mL
PBS with 0.02% sodium azide
P15090
ALBP, Adipocyte fatty acid binding protein, Fatty acid binding protein 4
Chicken
IgY
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2167
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit