Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponectin Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129901

Catalog No. PA129901


Add to cart

Description

Description

PA1-29901 detects Adiponectin from rat, mouse, human samples. PA1-29901 has been successfully used in immunoprecipitation, and Western blot applications. The PA1-29901 immunogen is: E.coli expressed recombit full length protein (Rat). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Adipose cells produce and secrete numerous physiologically important proteins, such as Lipoprotein Lipase, Leptin, and Adipocyte Complement Related protein of 30 kDa, also know as Acrp30 or Adiponectin. Adiponectin is a circulating protein that is secreted exclusively by differentiated adipocytes. During adipocyte differentiation, Adiponectin mRNA is induced > 100 fold. Adiponmectin imporves the ability of insulin to suppress glucose production, at sub physiological levels, thereby linking adipose tissue to whole body glucose regulation. Adiponectin function appears to be regulated by phosphatidylinositol 3 kinase (PI3k) since Adiponectin secretion is blocked by pharmacologic inhibitors of this kinase. Adiponectin mRNA is signigicantly reduced in adipose tissue of obese patients with type 2 diabetes. The structural similarity of Adiponectin to TNF alpha suggests that Adiponectin may play a role in pathogenesis of insulin resistance in Type 2 diabetes. Adiponectin is implicated as a regulator of whole body energy homeostasis.
Specifications

Specifications

Adiponectin
Polyclonal
Unconjugated
ADIPOQ
Liquid
ADIPOQ
E.coli expressed recombit full length protein (Rat).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunoprecipitation, Western Blot
0.5 mg/mL
PBS with 0.5% BSA, 30% glycerol and 0.01% thimerosal; pH 7.2
Q15848, Q60994
Adipocyte, C1q and collagen domain-containing protein, 30 kDa adipocyte complement-related protein, ACRP30, Adipose most abundant gene transcript 1, a
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11450, 246253, 9370
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit