Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADO Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524556

Catalog No. PIPA524556

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Human thiol dioxygenases include cysteine dioxygenase (CDO; MIM 603943) and cysteamine (2-amino ethane thiol) dioxygenase (ADO; EC 1.13.11.19). CDO adds 2 oxygen atoms to free cysteine, whereas ADO adds 2 oxygen atoms to free cysteamine to form hypotaurine.
Specifications

Specifications

ADO
Polyclonal
PBS with 0.09% sodium azide
Q96SZ5, Q6PDY2
C10orf22, 2-aminoethanethiol dioxygenase, cysteamine (2-aminoethanethiol) dioxygenase (ADO), cysteamine dioxygenase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
211488, 84890
Flow Cytometry, Western Blot
Unconjugated
ADO
Liquid
ADO
KLH conjugated synthetic peptide between 206-233 amino acids from the C-terminal region of human ADO
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit