Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Adropin, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126387H

Catalog No. NBP126387H


Add to cart

Description

Description

Adropin Polyclonal antibody specifically detects Adropin in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Adropin
HRP
PBS with No Preservative
Affinity Purified
ENHO
Synthetic peptide made to an internal portion of human Adropin (within residues 10-60). [Swiss-Prot# Q6UWT2]
8 kDa
0.1 ml
Diabetes Research, Lipid and Metabolism
Primary
375704
Human, Mouse
Western Blot
Western Blot
Q6UWT2
adropin, C9orf165, chromosome 9 open reading frame 165, energy homeostasis associated, Energy homeostasis-associated protein, GAAI470, UNQ470
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human and mouse. 88% sequence identity with bovine protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only