Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adropin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP126387SS
Description
Adropin Polyclonal specifically detects Adropin in Human, Mouse samples. It is validated for Western Blot.Specifications
Adropin | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
adropin, C9orf165, chromosome 9 open reading frame 165, energy homeostasis associated, Energy homeostasis-associated protein, GAAI470, UNQ470 | |
Rabbit | |
8 kDa | |
0.025 mL | |
Diabetes Research, Lipid and Metabolism | |
375704 | |
Human, Mouse | |
IgG |
Western Blot | |
1.11 mg/mL | |
Western Blot 2 ug/ml | |
Q6UWT2 | |
ENHO | |
Synthetic peptide made to an internal portion of human Adropin (within residues 10-60). [Swiss-Prot# Q6UWT2] | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. 88% sequence identity with bovine protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction