Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Advanced Glycated End Products, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP235353

Catalog No. NBP235353


Add to cart

Description

Description

Advanced Glycated End Products Polyclonal antibody specifically detects Advanced Glycated End Products in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Advanced Glycated End Products
0.2 mg/mL
Western Blot 1:100-1:400, ELISA 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:100-1:500
Affinity Purified
Native protein, Advanced Glycation End Product (AGE), conjugated to BSA (NPB353Ge91)
Immunogen affinity purified
RUO
Primary
Recognizes Advanced Glycation End Product (AGE)
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS (pH 7.4) and 50% Glycerol with 0.02% Sodium Azide
Rabbit
IgG
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only