Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLLT10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528399

Catalog No. PIPA528399


Add to cart

Description

Description

PA5-28399 targets AF10 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28399 immunogen is synthetic peptide corresponding to a region within amino acids 965 and 1027 of Human AF10. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a transcription factor and has been identified as a partner gene involved in several chromosomal rearrangements resulting in various leukemias. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

MLLT10
Polyclonal
Unconjugated
MLLT10
Liquid
MLLT10
Synthetic peptide corresponding to a region within amino acids 965 and 1027 of Human AF10
Protein A
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
7.29 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P55197
AF10, ALL1-fused gene from chromosome 10 protein, protein AF-10, type I AF10 protein, type III AF10 protein, type IV AF10 protein
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
8028
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit