Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-AF4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128728

Catalog No. NBP128728


Add to cart

Description

Description

AF4 Polyclonal antibody specifically detects AF4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative).
Specifications

Specifications

AF4
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:500 - 1:2000
P51825
AF-4, AF4/FMR2 family member 1, AF4/FMR2 family, member 1, AF4myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 2, ALL1-fused gene from chromosome 4 protein, FEL, MLLT2MGC134969, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 2, PBM1, pre-B-cell monocytic leukemia partner 1, Protein AF-4, Protein FEL, Proto-oncogene AF4
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Crab-eating macaque, African elephant and Northern white-cheeked gibbon.
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
AFF1
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human proto-oncogene AF4 using the numbering given in entry NP_005926.1 (GeneID 4299).
Immunogen affinity purified
RUO
Primary
4299
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only