Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AF6 Mouse, Unlabeled, Clone: 35, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610733

 View more versions of this product

Catalog No. BDB610733


Add to cart

Description

Description

AF6 (ALL-1 Fusion partner in chromosome 6) was discovered as the fusion partner of ALL-1. These genes undergo translocation and fusion in acute myeloid leukemias. AF6, or p180, was also identified as a Ras-interacting protein isolated from bovine brain. AF6 binds to the GTP-Ras but not to the inactive GDP, or ineffective forms of Ras. The interaction between AF6 and Ras may regulate the Ras signaling pathway since this association was prevented by the binding of c-Raf-1 to Ras. The Ras-binding site is located within amino acids 36-206 of AF6. AF6 is 1612 amino acids in length with homology to the DLG family of proteins located in cell-cell junctions, suggesting AF6 might be important in cell-to-cell communicative events. Furthermore, AF6 has the GLGF motif characteristic of DLG proteins that include Drosophilia's Canoe, implicated in the developmental pathway triggered by Notch activation. Like AF6, Canoe was also identified as a Ras-binding protein. The amino acid sequence and biochemical property similarities between Canoe and AF6 indicate that the latter may be important in developmental processes regulated by Ras.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

AF6
35
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
ALL-1 Fusion partner in chromosome 6
Affinity Purified
Human AF6 aa. 1091-1233
150μg
Cell Biology
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.