Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFMID Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526183

Catalog No. PIPA526183


Add to cart

Specifications

Specifications

AFMID
Polyclonal
PBS with 0.09% sodium azide
Q63HM1, Q8K4H1
KF, kynurenine formamidase, probable arylformamidase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
125061, 71562
Flow Cytometry, Western Blot
Unconjugated
AFMID
Liquid
AFMID
KLH conjugated synthetic peptide between 57-83 amino acids from the N-terminal region of human AFMID
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit