Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFTPH Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525674

Catalog No. PIPA525674

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

AFTPH may play a role in membrane trafficking.
Specifications

Specifications

AFTPH
Polyclonal
PBS with 0.09% sodium azide
Q6ULP2
Nbla10388, aftiphilin protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54812
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
AFTPH
Liquid
AFTPH
KLH conjugated synthetic peptide between 844-871 amino acids from the C-terminal region of human AFTPH
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit