Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENTG1 Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523322

Catalog No. PIPA523322


Add to cart

Description

Description

AGAP2 (also known as PIKE-A), a putative oncogene, shows elevated expression in cancer cells compared to non-transformed cells and has been implicated as regulators of cancer cell invasion into normal tissues. A contributor to brain, eye, breast and liver cancers, AGAP2 belongs to the centaurin gamma-like family with two ANK repeats, an Arf-GAP domain, a Miro domain and a PH domain is a GTPase-activating protein (GAP) for ARF1 and ARF5, which also shows strong GTPase activity. The Arf-GAP domain interacts with G domain and may regulate its GTPase activity. GAP activity of AGAP2 is stimulated by phosphatidylinositol 4,5-bisphosphate (PIP2) and, to a lesser extent, by phosphatidylinositol 3,4,5-trisphosphate (PIP3). Phosphatidic acid potentiates PIP2 stimulation. Several isoforms of AGAP2 have been reported.
Specifications

Specifications

CENTG1
Polyclonal
Unconjugated
AGAP2
Liquid
AGAP2
Synthetic peptide (34 ERVDDPELQDSI 45) of AGAP2 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Non-human Primate
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q99490
CENTG1, GGAP2, PIKE, Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 2, GTP-binding and GTPase activating protein 2, arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2, centaurin, gamma 1, cnt-g1, phosphatidylinositol-3-kinase enhancer, phosphoinositide 3-kinase enhancer
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
116986
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit