Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP3 Goat anti-Human, Mouse, Rat, Hamster, Monkey, Rabbit, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP140081

Catalog No. NBP140081


Add to cart

Description

Description

AGAP3 Polyclonal antibody specifically detects AGAP3 in Human, Mouse, Rat, Hamster, Monkey, Rabbit samples. It is validated for ELISA, Immunohistochemistry (Paraffin).
Specifications

Specifications

AGAP3
0.5 mg/mL
ELISA 1:32000, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml
Affinity Purified
AGAP3
Peptide with sequence C-NPSAELHRSPSLL, from the C Terminus of the protein sequence according to NP_114152.3.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Hamster, Human, Monkey, Mouse, Rabbit, Rat
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
TBS (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
ANK repeat and PH domain-containing protein 3, ArfGAP with GTPase domain, ankyrin repeat and PH domain 3, centaurin-gamma-3, Cnt-g3, CRAG, CRAM-associated GTPase, CRMP (collapsin response mediator protein) associated, FLJ16146, FLJ34452, gamma 3, MR1-interacting protein, MRIP-1
Goat
IgG
0.05 mg
Signal Transduction
Primary
116988
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only