Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-AGER, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP142612

Catalog No. NBP142612

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

AGER
Unconjugated
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3. with 0.05mg Thimerosal
Affinity Purified
AGER
A synthetic peptide corresponding to a sequence mapping at the N-terminal of human advanced glycosylation end-product-specific receptor(RAGE), identical to the related rat and mouse sequence.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute with 0.1 ml distilled water to desired concentration.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot 1-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 0.5-1 ug/ml
Q15109
advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products
Rabbit
IgG
0.1 mg
Cancer
Primary
177
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only