Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan G3, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA11745

Catalog No. PA1-1745

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Aggrecan is a member of large, aggregating proteoglycans (also including, versican, brevican and neurocan) found in articular cartilage. Aggrecan is composed of three major domains: G1, G2, and G3. Between the G1 and G2 domains there is an interglobulin region (IGD). The IGD region is the major site of cleavage by specific proteases like metalloproteinases (MMP’s) and aggrecanase. Aggrecan cleavage has been associated with a number of degenerative diseases including rheumatoid arthritis and osteoarthritis.The G3 domain is a complicated region produced by alternative splicing of exons in post-transcriptional processes. The human G3 domain consists of three modules and articular cartilage contains a mixture of the alternatively spliced G3 modules. G3 has been shown to facilitate glycosaminoglycan (GAG) attachment and overall aggrecan processing. The G3 domain is central to aggrecan turnover and function in cartilage.

PA1-1745 detects human, rat, mouse, bovine, and canine aggrecan G3.PA1-1745 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~350kDa protein representing the aggrecan G3 domain on the full length core.The PA1-1745 immunogen is a synthetic peptide corresponding to residues C D(2277) G H P M Q F E N W R P N Q P D N(2293) of human aggrecan.. This antibody is also referred to as anti-G3.
Specifications

Specifications

Aggrecan G3
Polyclonal
Unconjugated
ACAN
Purified
ACAN
Synthetic peptide corresponding to residues C D(2277) G H P M Q F E N W R P N Q P D N(2293) of human aggrecan.
Affinity chromatography
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
176
Western Blot
1mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P13608, Q28343, P16112, Q61282, P07897
AGC1, AGCAN, CSPG1, CSPGCP, MSK16, SEDK
Rabbit
IgG
100μg
Antibody
Polyclonal
Bovine, Canine, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit