Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ago2 Mouse anti-Human, Clone: 9E8.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04642
Description
Anti-Ago2 Antibody, clone 9E8.2 is a Mouse Monoclonal Antibody for detection of Ago2 also known as Argonaute-2, Eukaryotic translation initiation factor 2C and has been validated in western blotting and has been demonstrated to perform in Chimmunoprecipitation and ChIP-seq.Specifications
Ago2 | |
Monoclonal | |
Unconjugated | |
NM_012154; 27161 | |
Mouse | |
Immunoaffinity purified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Western Blot | |
9E8.2 | |
Ascites with 0.05% sodium azide | |
Ago-2, EIF2C2 | |
KLH-conjugated, synthetic peptide containing the sequence YSGAGPALAPPAPPPPIQG | |
100 μL | |
Human | |
Ascites |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only