Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGPAT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527616

Catalog No. PIPA527616


Add to cart

Description

Description

PA5-27616 targets AGPAT1 in WB applications and shows reactivity with Human samples. The PA5-27616 immunogen is recombit fragment corresponding to a region within amino acids 106 and 283 of Human AGPAT1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes an enzyme that converts lysophosphatidic acid (LPA) into phosphatidic acid (PA). LPA and PA are two phospholipids involved in signal transduction and in lipid biosynthesis in cells. This enzyme localizes to the endoplasmic reticulum. This gene is located in the class III region of the human major histocompatibility complex. Alternative splicing results in two transcript variants encoding the same protein.
Specifications

Specifications

AGPAT1
Polyclonal
Unconjugated
AGPAT1
Liquid
AGPAT1
Recombit fragment corresponding to a region within amino acids 106 and 283 of Human AGPAT1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q99943
1-AGPAT1, G15, LPAAT-alpha, LPAATA, 1-AGP acyltransferase 1, 1-AGPAT 1, 1-acyl-sn-glycerol-3-phosphate acyltransferase alpha, 1-acylglycerol-3-phosphate O-acyltransferase 1 (acetoacetly Coenzyme A thiolase), lysophosphatidic acid acyltransferase alpha, lysophospholipid acyltransferase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10554
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit