Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGR3 Mouse anti-Human, Clone: AGR3.2, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA110049
Description
This antibody recognizes the epitope QYSQALKKV within AGR3.
AGR3 (Anterior Gradient 3) protein, also known as AG3 (hAG-3, HAG3 in human), or BCMP11, is a secreted cytoplasmic protein which is involved in metastasis induction and p53 tumour supressor inhibition. It may serve as molecular marker and potential therapeutic target for hormone-responsive breast tumours. Its Xenopus homolog is associated with anteroposterior fate determination during early development.Specifications
AGR3 | |
Monoclonal | |
Unconjugated | |
Q8TD06 | |
AGR3 | |
Purified human AGR3 protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
AGR3.2 | |
AGR3 | |
UNQ642/PRO1272, AG3, BCMP11, HAG3, PDIA18, hAG-3, AG-3, anterior gradient protein 3 homolog, breast cancer membrane protein 11, protein disulfide isomerase family A, member 18 | |
Mouse | |
Protein A | |
RUO | |
155465 | |
4° C, do not freeze | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title