Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGXT2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526833

Catalog No. PIPA526833


Add to cart

Description

Description

The protein encoded by this gene is a class III pyridoxal-phosphate-dependent mitochondrial aminotransferase. It catalyzes the conversion of glyoxylate to glycine using L-alanine as the amino donor.
Specifications

Specifications

AGXT2
Polyclonal
PBS with 0.09% sodium azide
Q9BYV1
AGT2, DAIBAT, (R)-3-amino-2-methylpropionate--pyruvate transaminase, alanine--glyoxylate aminotransferase 2, mitochondrial, beta-ALAAT II, beta-alanine-pyruvate aminotransferase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64902
Flow Cytometry, Western Blot
Unconjugated
AGXT2
Liquid
AGXT2
KLH conjugated synthetic peptide between 437-463 amino acids from the C-terminal region of human AGXT2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit