Learn More
AHCY Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA527400
Catalog No. PIPA527400
Add to cart
Description
Description
PA5-27400 targets AHCY in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-27400 immunogen is recombit fragment corresponding to a region within amino acids 168 and 419 of Human AHCY. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
S-adenosylhomocysteine hydrolase catalyzes the reversible hydrolysis of S-adenosylhomocysteine (AdoHcy) to adenosine (Ado) and L-homocysteine (Hcy). Thus, it regulates the intracellular S-adenosylhomocysteine (SAH) concentration thought to be important for transmethylation reactions. Deficiency in this protein is one of the different causes of hypermethioninemia. S-adenosylhomocysteine hydrolase belongs to the adenosylhomocysteinase family.
Specifications
Specifications
AHCY
Polyclonal
Unconjugated
AHCY
Liquid
AHCY
Recombit fragment corresponding to a region within amino acids 168 and 419 of Human AHCY
Protein A
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
8.5 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P23526
SAHH, S-adenosyl-L-homocysteine hydrolase, S-adenosylhomocysteine hydrolase, adoHcyase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
191