Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-AIF, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP177302

Catalog No. NBP177302

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

AIF
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
O95831
AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
At least five isoforms of AIF are known to exist; this antibody will detect all isoforms except isoform 5.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Ion Exchange Chromatography
AIFM1
Antibody was raised against a 15 amino acid peptide near the center of human AIF. The immunogen is located within amino acids 500 - 550 of AIF.
71 kDa
0.1 mg
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Mitochondrial Markers, Tumor Suppressors
Primary
9131
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only