Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMID Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524562

Catalog No. PIPA524562

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and mouse based on sequence homology.

AIFM2 is significant homology to NADH oxidoreductases and the apoptosis-inducing factor PDCD8/AIF. The protein has been shown to induce apoptosis. This protein is found to be induced by tumor suppressor protein p53 in colon caner cells.
Specifications

Specifications

AMID
Polyclonal
PBS with 0.09% sodium azide
Q9BRQ8, Q8BUE4
AMID, PRG3, RP11-367H5.2, apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death, apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death, apoptosis-inducing factor 2, p53-responsive gene 3 protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
71361, 84883
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
AIFM2
Liquid
AIFM2
KLH conjugated synthetic peptide between 319-348 amino acids from the C-terminal region of human AIFM2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit