Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aiolos Mouse anti-Mouse, PE, Clone: S48-791, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564874

Catalog No. BDB564874


Add to cart

Description

Description

The S48-791 monoclonal antibody specifically recognizes the transcription factor, Aiolos, which is encoded by Ikzf3 , IKAROS family zinc finger 3. Aiolos belongs to the Ikaros family of zinc-finger proteins that includes Ikaros and Helios. Aiolos can form homodimers or heterodimers with other Ikaros family members. Aiolos is expressed in committed lymphoid progenitors, thymocytes, and mature types of T and B lymphocytes and NK cells. It is not expressed by hematopoietic stem cells. Aiolos plays an important role in the control of B lymphocyte differentiation and proliferation and is specifically required for the generation of long-lived, high affinity plasma cells in the bone marrow. Aiolos is involved in regulating BCL2 expression and controlling apoptosis in T-cells in an IL2-dependent manner. Aiolos is also expressed by Th17 cells and reportedly promotes their differentiation by downregulating IL-2 expression.

Specifications

Specifications

Aiolos
S48-791
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Ikzf3; IKAROS family zinc finger 3; Zfpn1a3; Znfn1a3
Affinity Purified
Human Aiolos Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit