Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
adenylate kinase 3, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human AK3 protein.
Manufacturer: Abnova Corporation H00050808B01
Description
Sequence: MGASARLLRAVIMGAPGSGKGTVSSRITTHFELKHLSSGDLLRDNMLRGTEIGVLAKAFIDQGKLIPDDVMTRLALHELKNLTQYSWLLDGFPRTLPQAEALDRAYQIDTVINLNVPFEVIKQRLTARWIHPASGRVYNIEFNPPKTVGIDDLTGEPLIQREDDKPETVIKRLKAYEDQTKPVLEYYQKKGVLETFSGTETNKIWPYVYAFLQTKVPQRSQKASVTPSpecifications
adenylate kinase 3 | |
Unconjugated | |
No additive | |
NM_016282.2 | |
AK3L1|AK6|AKL3L|AKL3L1|FIX | |
Mouse | |
50 uL | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
50808 |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human AK3 protein. | |
AK3 | |
Serum | |
AK3 | |
AK3 (NP_057366.2, 1 a.a. ~ 227 a.a) full-length human protein. | |
RUO | |
Y | |
Polyclonal | |
Human, Murine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title