Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP12 Sheep anti-Rat, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA124645

Catalog No. PIPA124645


Add to cart

Description

Description

PA1-24645 detects AKAP12 from rat samples. PA1-24645 has been successfully used in Western blot procedures. The PA1-24645 immunogen is purified, full length, native rat protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

AKAP12
Polyclonal
Unconjugated
Akap12
Liquid
AKAP12
Purified, full length, native rat protein.
Protein A
RUO
Antibody
Polyclonal
Rat
Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.08% sodium azide
Q5QD51
AKAP12A, AKAP12B, AKAP12G
Sheep
IgG
125 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
83425
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit