Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP149 Mouse, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610720

Catalog No. BDB610720


Add to cart

Description

Description

The targeting of protein kinases and phosphatases to cellular compartments in a timely manner may provide specificity in signal transduction events. For instance, attachment of type II cAMP-dependent Protein Kinase (PKA) to the cytoskeleton occurs through the binding of the regulatory subunit (RII) to microtubule associated-protein 2. Several proteins, such as microtubule-associated protein 2, have been identified as PKA type I/II anchoring proteins and form a family named AKAP (A-Kinase Anchor

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

AKAP149
6
Unconjugated
Affinity Purified
Human AKAP149 aa. 66-212
50μg
Cell Biology
Primary
Human, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit