Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-AKAP9, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP147205

Catalog No. NBP147205


Add to cart

Description

Description

AKAP9 Polyclonal antibody specifically detects AKAP9 in Human, Canine samples. It is validated for Peptide ELISA.
Specifications

Specifications

AKAP9
0.5 mg/mL
Peptide ELISA Detection limit 1:64000
NP_005742.4
A kinase (PRKA) anchor protein (yotiao) 9, AKAP 350, AKAP 450, AKAP350A-kinase anchor protein 450 kDa, AKAP450A-kinase anchor protein 350 kDa, AKAP-9, Centrosome- and Golgi-localized PKN-associated protein, CG-NAPAKAP 120-like protein, hgAKAP 350, HYPERION, KIAA0803A-kinase anchor protein 9, MU-RMS-40.16A, PRKA9AKAP9-BRAF fusion protein, Protein hyperion, protein kinase A anchoring protein 9, Protein kinase A-anchoring protein 9, Protein yotiao, YOTIAOkinase N-associated protein
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize isoforms 2 and 3 (NP_005742.4; NP_671714.1)
ELISA
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
AKAP9
Peptide with sequence QRKAQSDGQSPSKK-C corresponding to N-Terminus according to NP_005742.4, NP_671714.1.
Immunogen affinity purified
RUO
Primary
10142
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only