Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP95 Mouse anti-Human, Unlabeled, Clone: 47, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610994

Catalog No. BDB610994


Add to cart

Description

Description

Induction of cAMP results in the activation of the cAMP-dependent protein kinase (PKA) which, in turn, phosphorylates and regulates downstream effectors. The localization and, hence, the specificity of PKA action is mediated by the interaction of four different PKA regulatory subunits (RIα, RIβ, RIIα, and RIIβ) and specific PKA anchoring proteins. Several proteins have been identified as PKA type I/II anchoring proteins and form a family named AKAP (A-nchor Proteins). Several AKAPs (AKAP79/75, AKAP350, and AKAP250) associate with PKA type II and direct its localization to cortical actin, centrosomes, and filopodia. AKAP95 is a specific nuclear anchoring protein that contains two zinc finger motifs and an RII binding domain. Although AKAP95 does not interact with RIIβ, it has a high affinity for RIIα. During interphase, AKAP95 is bound to DNA. However, entry into mitosis results in AKAP95 detachment from the DNA and association with RIIα. Although the specific function of this AKAP95/RIIα heterodimer is unknown, its formation is cell cycle-dependent and may be essential for proper progression through the cell cycle.

Specifications

Specifications

AKAP95
47
Unconjugated
Affinity Purified
Human AKAP95 aa. 460-781
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit