Learn More
MilliporeSigma™ anti-AKR1B10 Clone: 1A6, Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ ST1507100UG
Catalog No. ST1507100UG
Add to cart
Description
Description
Specifically detects AKR1B10 Clone: 1A6 in Human samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immuno Blotting
This gene product encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member can efficiently reduce aliphatic and aromatic aldehydes, and it is less active on hexoses. It is highly expressed in adrenal gland, small intestine, and colon, and may play an important role in liver carcinogenesis.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
AKR1B10
1A6
In PBS, pH 7.2.
Murine
IgG2a
RUO
Primary
Recognizes the ∽37kDa AKR1B10 protein in HepG2 and HeLa cells and human colon tissue.
ELISA, Immuno Blotting, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Purified
A recombinant polypeptide corresponding to amino acids 76-144 of human AKR1B10, expressed as a GST fusion protein
100μg
−20°C
Monoclonal
Human