Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1B10, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523017

Catalog No. PIPA523017

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

AKR1B10, also known as Aldo-keto reductase family 1, member B10, AKR1B10 is a monomeric protein that efficiently catalyzes the reduction of aromatic and aliphatic aldehydes and ketones. AKR1B10 is ubiquitously expressed in many human tissues but is highly expressed in small intestine, colon and adrenal gland. This protein is pathogenically involved in diabetic complications and has been reported that AKR1B10 is overexpressed in human tumors, such as liver, breast, and lung cancer, and may play a critical role in the development and progression of cancer.

This antibody does not cross react with mouse, rat or rabbit.
Specifications

Specifications

AKR1B10
Polyclonal
Unconjugated
AKR1B10
Purified
AKR1B10
A highly specific 15 amino acid portion of human AKR1B10 (within amino acids 100-150)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1mg/mL
tris HCl with 100mM NaCl, 50% glycerol and no preservative
O60218
AKR1B11, AKR1B12, ALDRLn, ARL-1, ARL1, HIS, HSI, ARP, SI reductase, aldo-keto reductase family 1 member B10, aldo-keto reductase family 1, member B11 (aldose reductase-like), aldose reductase-like 1, aldose reductase-like peptide, aldose reductase-related protein, hARP, small intestine reductase
Rabbit
IgG
100μL
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
57016
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit