Learn More
AKR1C2 (M03), Mouse anti-Human, Clone: 3C11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant AKR1C2.
Supplier: Abnova Corporation H00001646M03
Description
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols using NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme binds bile acid with high affinity, and shows minimal 3-alpha-hydroxysteroid dehydrogenase activity. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14. [provided by RefSeq
Sequence: EEPWVDPNSPVLLEDPVLCALAKKHKRTPALIALRYQLQRGVVVLAKSYNEQRIRQNVQVFEFQLTSEEMKAIDGLNRNVRYLTLDIFAGPPNYPVSDEYSpecifications
AKR1C2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC063574 | |
AKR1C2 | |
AKR1C2 (AAH63574, 224 a.a. ∼ 323 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
3C11 | |
Mouse monoclonal antibody raised against a partial recombinant AKR1C2. | |
AKR1C2 | |
AKR1C-pseudo/BABP/DD/DD2/DDH2/HAKRD/HBAB/MCDR2 | |
Mouse | |
Affinity chromatography | |
RUO | |
1646 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.