Learn More
Akt Mouse, Unlabeled, Clone: 2, BD
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 610876
Description
Protein Kinase B (PKB), also known as Rac (Related to the A and C kinases) Protein Kinase, shows 73% and 68% to Protein Kinase C (PKC) and Protein Kinase A (PKA), respectively. It was also identified as the product of the v-akt oncogene from T-cell lymphomas. Akt/PKB mRNA has been reported to be expressed in a wide variety of cell lines and has been detected in porcine brain, heart, liver, kidney, muscle, and ovary. Rac phosphorylates a number of physiological substrates including MBP, glycogen synthetase, PKA RII subunit, and histone H1. Akt/PKB is activated in response to growth factors through the activation of PI3-kinase and Ras. Activation of PI3-Kinase generates phosphatidylinositol 3,4 bisphosphate which may induce the membrane translocation of Akt/PKB coincident with its phosphorylation and activation. Cellular stress leading to the activation of the p38 MAP kinase also induces Akt/PKB activation, indicating a multiplicity of signaling pathways that activate Akt/PKB. Upon activation, Akt/PKB associates with some members of the PKC family of kinases, such as PKCδ and PKCζ. Downstream of Akt/PKB, GSK3 is thus far the only identified substrate for Akt/PKB phosphorylated in vivo.
Immunofluorescence, Western Blotting
Specifications
Akt | |
Monoclonal | |
250μg/mL | |
PKBα; RAC-PK | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
IgG2a |
Western Blot | |
2 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human PKBα/Akt1 aa. 1-480 | |
50 μg | |
Cell Biology | |
Canine, Drosophila, Human, Murine, Rat |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.