Learn More
Akt/PKB Mouse anti-Human, Mouse, Rat, Clone: SKB1, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05591MG
Catalog No. 05-591-MGMI
Add to cart
Description
Description
This Anti-Akt/PKB Antibody, PH Domain, clone SKB1 is validated for use in immunoprecipitation, western blotting for the detection of Akt/PKB.
Specifications
Specifications
Akt/PKB
SKB1
Protein G Purified immunoglobulin in 0.01M Phosphate Buffer containing no preservative, pH 7.1.
Purified
Murine
IgG
1mg
Primary
Human, Murine, Rat
Immunoprecipitation, Western Blot
Unconjugated
NM_005163
AKT1, RAC, PRKBA, MGC99656, RAC-ALPHA, RAC-PK-alpha, C-AKT, PKB, AKT
GST-fusion protein corresponding to residues 1-149 of human Akt
Protein G purfied
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Monoclonal