Learn More
MilliporeSigma™ Akt/PKB Mouse anti-Human, Mouse, Rat, Clone: SKB1
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ 05591
Catalog No. 05-591-MI
Add to cart
Description
Description
Detect Akt/PKB using this Anti-Akt/PKB Antibody, PH Domain, clone SKB1 validated for use in flow cytometry, immunocytochemistry, immunoprecipitation and western blotting.
Specifications
Specifications
Akt/PKB
SKB1
Protein G Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 0.02Mphosphate buffer, pH 7.6, 0.25M NaCl, 0.1% sodium azide and 30% glycerol.
Purified
Murine
IgG
100μg
Primary
Human, Murine, Rat
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_005163
AKT1, RAC, PRKBA, MGC99656, RAC-ALPHA, RAC-PK-alpha, C-AKT, PKB, AKT
GST-fusion protein corresponding to residues 1-149 of human Akt1
Protein G purfied
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Monoclonal